1-Azido-1-deoxy-β-D-lactopyranoside 97%

Supplier: AFG BIOSCIENCE LLC

C51555-1G C51555-250MG C51555-500MG
76619-574EA 1024.85 CAD
76619-574 76619-570 76619-572
1-Azido-1-deoxy-β-D-lactopyranoside 97%
1-Azido-1-deoxy-β-D-lactopyranoside
Formula: C₁₂H₂₁N₃O₁₀
MW: 367.31 g/mol
Storage Temperature: Freezer
MDL Number: MFCD05664202
CAS Number: 69266-16-6

Order Now

Specification Test Results

Purity Limit 97%
Appearance White powder


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR