6-Azido-6-deoxy-D-mannose 95%

Supplier: AFG Bioscience

C51543-100MG C51543-25MG C51543-50MG
76619-558EA 1323.99 CAD
76619-558 76619-554 76619-556
6-Azido-6-deoxy-D-mannose 95%
6-Azido-6-deoxy-D-mannose
Formula: C₆H₁₁N₃O₅
MW: 205.17 g/mol
Storage Temperature: Freezer
CAS Number: 316379-15-4

Order Now

Specification Test Results

Purity Limit 95%
Appearance light-yellow sticky oil


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR