6-Azido-6-deoxy-D-glucose peracetate ≥95%

Supplier: AFG Bioscience

Synonyms: 1,2,3,4-Tetra-O-acetyl-6-azido-6-deoxy-D-glucopyranose

C51538-1G C51538-500MG C51538-100MG C51538-250MG
76619-546EA 1342.57 CAD
76619-546 76619-544 77888-731 77888-732
6-Azido-6-deoxy-D-glucose peracetate ≥95%
6-Azido-6-deoxy-D-glucose peracetate
Formula: C₁₄H₁₉N₃O₉
MW: 373.32 g/mol
Storage Temperature: Refrigerator
CAS Number: 189618-61-9

Order Now

Specification Test Results

Purity Limit > 95%
Appearance White Powder


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR