2-acetylamino-6-azido-2,6-dideoxy-D-glucose ≥95%

Supplier: AFG Bioscience

C51535-100MG C51535-50MG
76619-530EA 1012.46 CAD
76619-530 76619-528
2-acetylamino-6-azido-2,6-dideoxy-D-glucose ≥95%
2-acetylamino-6-azido-2,6-dideoxy-D-glucose
Formula: C₈H₁₄N₄O₅
Storage Temperature: Freezer
CAS Number: 219814-62-7

Order Now

Specification Test Results

Purity Limit > 95%
Appearance White Powder


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR