2-Azido-2-deoxy-D-glucose 97%

Supplier: AFG Bioscience

C51531-1G C51531-2G C51531-500MG
76619-516EA 474.35 CAD
76619-516 76619-518 76619-514
2-Azido-2-deoxy-D-glucose 97%
2-Azido-2-deoxy-D-glucose
Formula: C₆H₁₁N₃O₅
MW: 205.17 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09750677
CAS Number: 56883-39-7

Order Now

Specification Test Results

Purity Limit 97%
Appearance White Crystalline Solid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR