2-acetylamino-6-azido-2,6-dideoxy-D-galactose, peracetate ≥95%

Supplier: AFG BIOSCIENCE LLC

Synonyms: 1,3,4 -Tri-O-acetyl-2-acetylamino-6-azido-2,6-dideoxy-D-galactose

C51530-100MG C51530-50MG
76619-512EA 700.94 CAD
76619-512 76619-510
2-acetylamino-6-azido-2,6-dideoxy-D-galactose, peracetate ≥95%
2-acetylamino-6-azido-2,6-dideoxy-D-galactose, peracetate
Formula: C₁₄H₂₀N₄O₈
MW: 372.33 g/mol
Storage Temperature: Refrigerator
CAS Number: 657363-19-4

Order Now

Specification Test Results

Purity Limit > 95%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR