1-Azido-1-deoxy-β-D-galactopyranoside tetraacetate 97%

Supplier: AFG Bioscience

C51527-1G C51527-2G C51527-5G
76619-500EA 237.36 CAD
76619-500 76619-502 76619-504
1-Azido-1-deoxy-β-D-galactopyranoside tetraacetate 97%
1-Azido-1-deoxy-β-D-galactopyranoside tetraacetate
Formula: C₁₄H₁₉N₃O₉
Storage Temperature: Refrigerator
MDL Number: MFCD00181721
CAS Number: 13992-26-2

Order Now

Specification Test Results

Purity Limit 97%
Appearance White Crystalline Solid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR