2-acetylamino-6-azido-2,6-dideoxy-D-galactose ≥95%

Supplier: AFG BIOSCIENCE LLC

C51525-100MG C51525-50MG
76619-492EA 1012.46 CAD
76619-492 76619-490
2-acetylamino-6-azido-2,6-dideoxy-D-galactose ≥95%
2-acetylamino-6-azido-2,6-dideoxy-D-galactose
Formula: C₈H₁₄N₄O₅
MW: 246.22 g/mol
Storage Temperature: Freezer
CAS Number: 1190619-44-3

Order Now

Specification Test Results

Purity Limit > 95%
Appearance White Powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR