N-azidoacetylmannosamine peracetate 95%

Supplier: AFG Bioscience

Synonyms: Ac4ManNAz

C51520-100MG C51520-50MG C51520-25MG
76619-472EA 864.13 CAD
76619-472 76619-470 77888-719
N-azidoacetylmannosamine peracetate 95%
N-azidoacetylmannosamine peracetate
Formula: C₁₆H₂₂N₄O₁₀
MW: 430.37 g/mol
Storage Temperature: Freezer
CAS Number: 361154-30-5

Order Now

Specification Test Results

Purity Limit 95%
Appearance white powder


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR