N-azidoacetylmannosamine 95%
Supplier: AFG Bioscience
C51519-100MG
C51519-50MG
C51519-10MG
C51519-25MG
76619-468EA
1570.03
CAD
76619-468
76619-466
77888-717
77888-718
N-azidoacetylmannosamine 95%
N-azidoacetylmannosamine
Formula:
C₈H₁₄N₄O₆ Storage Temperature: Freezer |
CAS Number:
361154-23-6 |
Specification Test Results
Purity Limit | 95% |
Appearance | white powder |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...