N-azidoacetylgalactosamine peracetate 95%

Supplier: AFG Bioscience

C51518-100MG C51518-50MG C51518-25MG
76619-464EA 870.33 CAD
76619-464 76619-462 77888-716
N-azidoacetylgalactosamine peracetate 95%
N-azidoacetylgalactosamine peracetate
Formula: C₁₆H₂₂N₄O₁₀
Storage Temperature: Freezer
CAS Number: 653600-56-7

Order Now

Specification Test Results

Purity Limit 95%
Appearance white powder


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR