N-azidoacetylgalactosamine 95%

Supplier: AFG BIOSCIENCE LLC

Synonyms: 2-[(2-Azidoacetyl)amino]-2-deoxy-D-galactose, GalNAz

C51517-100MG C51517-50MG
76619-460EA 1578.69 CAD
76619-460 76619-458
N-azidoacetylgalactosamine 95%
N-azidoacetylgalactosamine
Formula: C₈H₁₄N₄O₆
MW: 262.22 g/mol
Storage Temperature: Freezer
CAS Number: 869186-83-4

Order Now

Specification Test Results

Purity Limit 95%
Appearance white powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR