N-azidoacetylglucosamine peracetate 95%

Supplier: AFG Bioscience

Synonyms: Ac4GlcNAz

C51516-100MG C51516-50MG C51516-25MG
76619-456EA 869.08 CAD
76619-456 76619-454 77888-713
N-azidoacetylglucosamine peracetate 95%
N-azidoacetylglucosamine peracetate
Formula: C₁₆H₂₂N₄O₁₀
MW: 430.37 g/mol
Storage Temperature: Freezer
CAS Number: 98924-81-3

Order Now

Specification Test Results

Purity Limit 95%
Appearance white powder


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR