N-azidoacetylglucosamine 95%

Supplier: AFG Bioscience

Synonyms: GlcNAz

C51515-100MG C51515-50MG C51515-25MG C51515-10MG
76619-452EA 1576.22 CAD
76619-452 76619-450 77888-712 77888-711
N-azidoacetylglucosamine 95%
N-azidoacetylglucosamine
Formula: C₈H₁₄N₄O₆
Storage Temperature: Freezer
CAS Number: 92659-90-0

Order Now

Specification Test Results

Purity Limit 95%
Appearance Off-white powder


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR