2-Azidoethyl β-lactopyranoside 95%

Supplier: AFG Bioscience

C51514-100MG C51514-50MG
76619-448EA 506.23 CAD
76619-448 76619-446
2-Azidoethyl β-lactopyranoside 95%
2-Azidoethyl β-lactopyranoside
Formula: C₁₄H₂₅N₃O₁₁
Storage Temperature: Freezer
CAS Number: 230286-11-0

Order Now

Specification Test Results

Purity Limit 95%
Appearance Colorless Syrup


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR