2-Azidoethyl α-rhamnoside ≥95%

Supplier: AFG Bioscience

C51513-100MG C51513-50MG
76619-444EA 631.1 CAD
76619-444 76619-442
2-Azidoethyl α-rhamnoside ≥95%
2-Azidoethyl α-rhamnoside
Formula: C₈H₁₅N₃O₅
Storage Temperature: Freezer
CAS Number: 1381853-30-0

Order Now

Specification Test Results

Purity Limit > 95%
Appearance White Solid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR