2-azidoethyl α-D-Mannoside 95%

Supplier: AFG Bioscience

Synonyms: α-D-Man-Et-N3

C51512-100MG C51512-250MG C51512-500MG
76619-436EA 316.47 CAD
76619-436 76619-438 76619-440
2-azidoethyl α-D-Mannoside 95%
2-azidoethyl α-D-Mannoside
Formula: C₈H₁₅N₃O₆
MW: 249.22 g/mol
Storage Temperature: Refrigerator
CAS Number: 155196-97-7

Order Now

Specification Test Results

Purity Limit 95%
Appearance White Solid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR