2-Azidoethyl β-Galactopyranoside 95%

Supplier: AFG Bioscience

C51510-1G C51510-250MG C51510-500MG
76619-428EA 934.59 CAD
76619-428 76619-424 76619-426
2-Azidoethyl β-Galactopyranoside 95%
2-Azidoethyl β-Galactopyranoside
Formula: C₈H₁₅N₃O₆
Storage Temperature: Freezer
CAS Number: 151651-54-6

Order Now

Specification Test Results

Purity Limit 95%
Appearance Colorless Syrup


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR