2-azidoethyl α-D-Galactoside ≥95%
Supplier: AFG BIOSCIENCE LLC
C51509-100MG
C51509-50MG
76619-422EA
545.18
CAD
76619-422
76619-420
2-azidoethyl α-D-Galactoside ≥95%
2-azidoethyl α-D-Galactoside
Formula:
C₈H₁₅N₃O₆ MW: 249.22 g/mol Storage Temperature: Refrigerator |
CAS Number:
680988-10-7 |
Specification Test Results
Purity Limit | > 95% |
Appearance | Sticky Syrup |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...