α-Man-PEG3-Azide 95%

Supplier: AFG Bioscience

C51505-25MG C51505-50MG
76619-408EA 473.48 CAD
76619-408 76619-410
α-Man-PEG3-Azide 95%
α-Man-PEG3-Azide
Formula: C₁₂H₂₃N₃O₈
Storage Temperature: Freezer
CAS Number: 246855-76-5

Order Now

Specification Test Results

Purity Limit 95%
Appearance White powder


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR