β-Glc-PEG3-Azide 95%

Supplier: AFG BIOSCIENCE LLC

Synonyms: 2-{2-[2-azidoethoxy]ethoxy}ethyl β-D-glucopyranoside

C51504-25MG C51504-50MG
76619-404EA 473.48 CAD
76619-404 76619-406
β-Glc-PEG3-Azide 95%
β-Glc-PEG3-Azide
Formula: C₁₂H₂₃N₃O₈
MW: 337.33 g/mol
Storage Temperature: Freezer
CAS Number: 156058-83-2

Order Now

Specification Test Results

Purity Limit 95%
Appearance White powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR