β-Gal-PEG3-Azide ≥95%

Supplier: AFG BIOSCIENCE LLC

Synonyms: 2-{2-[2-azidoethoxy]ethoxy}ethyl β-D-galactopyranoside

C51503-25MG C51503-50MG
76619-400EA 395.6 CAD
76619-400 76619-402
β-Gal-PEG3-Azide ≥95%
β-Gal-PEG3-Azide
Formula: C₁₂H₂₃N₃O₈
MW: 337.33 g/mol
Storage Temperature: Freezer
CAS Number: 126765-27-3

Order Now

Specification Test Results

Purity Limit > 95%
Appearance White powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR