β-GlcNAc-PEG3-Azide ≥95%

Supplier: AFG Bioscience

C51502-25MG C51502-50MG
76619-396EA 389.41 CAD
76619-396 76619-398
β-GlcNAc-PEG3-Azide ≥95%
β-GlcNAc-PEG3-Azide
Formula: C₁₄H₂₆N₄O₈
Storage Temperature: Freezer
CAS Number: 86520-54-9

Order Now

Specification Test Results

Purity Limit > 95%
Appearance White powder


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR