Fmoc-Thr(Ac3GalN3α)-OtBu ≥95%

Supplier: AFG Bioscience

C51085-1G C51085-250MG C51085-500MG
76619-390EA 2373.62 CAD
76619-390 76619-386 76619-388
Fmoc-Thr(Ac3GalN3α)-OtBu ≥95%
Fmoc-Thr(Ac3GalN3α)-OtBu
Formula: C₃₅H₄₂N₄O₁₂
MW: 710.74 g/mol
Storage Temperature: Freezer
CAS Number: 12791-77-7

Order Now

Specification Test Results

Purity Limit > 95%
Appearance White Powder


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR