Fmoc-Ser(Ac3GalN3α)-OtBu ≥95%
Supplier: AFG Bioscience
Synonyms:
N-(9-Fluorenylmethoxycarbonyl)-O-(3,4,6-tri-O-acetyl-2-azido-2-desoxy-α-D-galactopyranosyl)-L-serin-tert-butylester
C51084-1G
C51084-250MG
C51084-500MG
76619-384EA
2373.62
CAD
76619-384
76619-380
76619-382
Fmoc-Ser(Ac3GalN3α)-OtBu ≥95%
Fmoc-Ser(Ac3GalN3α)-OtBu
Formula:
C₃₄H₄₀N₄O₁₂ MW: 696.7 g/mol Storage Temperature: Freezer |
CAS Number:
110797-36-9 |
Specification Test Results
Purity Limit | > 95% |
Appearance | White Powder |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...