2-Acetamido-2-deoxy-α-D-galactopyranosyl L-serine ≥95%
Supplier: AFG Bioscience
C51060-10MG
C51060-25MG
C51060-5MG
76619-358EA
1574.98
CAD
76619-358
76619-360
76619-356
2-Acetamido-2-deoxy-α-D-galactopyranosyl L-serine ≥95%
2-Acetamido-2-deoxy-α-D-galactopyranosyl L-serine
Formula:
C₁₁H₂₀N₂O₈ Storage Temperature: Freezer |
MDL Number:
MFCD01863462 CAS Number: 67262-86-6 |
Specification Test Results
Purity Limit | > 95% |
Appearance | White Powder |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...