Fmoc-L-Thr (α-D-Man(Ac)4)-OH 95%

Supplier: AFG BIOSCIENCE LLC

Synonyms: N-α-(9-Fluorenylmethyloxycarbonyl)-O-β-(2,3,4,6-tetra-O-acetyl-α-D-mannopyranosyl)-L-threonine

C51031-100MG C51031-250MG C51031-25MG
76619-346EA 1186.8 CAD
76619-346 76619-348 76619-344
Fmoc-L-Thr (α-D-Man(Ac)4)-OH 95%
Fmoc-L-Thr (α-D-Man(Ac)4)-OH
Formula: C₃₃H₃₇NO₁₄
MW: 671.65 g/mol
Storage Temperature: Freezer
CAS Number: 169219-08-3

Order Now

Specification Test Results

Purity Limit 95%
Appearance White powder


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR