Fmoc-L-Thr (β-D-Gal(Ac)4)-OH 95%

Supplier: AFG Bioscience

C51028-250MG C51028-25MG C51028-100MG
76619-336EA 1246.11 CAD
76619-336 76619-332 76619-334
Fmoc-L-Thr (β-D-Gal(Ac)4)-OH 95%
Fmoc-L-Thr (β-D-Gal(Ac)4)-OH
Formula: C₃₃H₃₇NO₁₄
Storage Temperature: Freezer
CAS Number: 127656-85-3

Order Now

Specification Test Results

Purity Limit 95%
Appearance White powder


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR