Fmoc-L-Ser(β-Xyl(Ac3))-OH 95%

Supplier: AFG Bioscience

C51016-100MG C51016-250MG C51016-25MG
76619-316EA 1186.8 CAD
76619-316 76619-318 76619-314
Fmoc-L-Ser(β-Xyl(Ac3))-OH 95%
Fmoc-L-Ser(β-Xyl(Ac3))-OH
Formula: C₂₉H₃₁NO₁₂
Storage Temperature: Freezer
MDL Number: MFCD05663658
CAS Number: 105678-33-9

Order Now

Specification Test Results

Purity Limit 95%
Appearance White powder


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR