Fmoc-L-Ser((Ac)3-α-Fuc)-OH ≥95%

Supplier: AFG Bioscience

Synonyms: N-α-(Fluorenylmethoxycarbonyl)-O-β-(2,3,4-tri-O-acetyl-α-L-fucopyranosyl)-L-serine

C51015-100MG C51015-250MG C51015-25MG
76619-310EA 1186.8 CAD
76619-310 76619-312 76619-308
Fmoc-L-Ser((Ac)3-α-Fuc)-OH ≥95%
Fmoc-L-Ser((Ac)3-α-Fuc)-OH
Formula: C₃₀H₃₃NO₁₂
MW: 599.58 g/mol
Storage Temperature: Freezer
CAS Number: 173935-46-1

Order Now

Specification Test Results

Purity Limit > 95%
Appearance White Powder


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR