Fmoc-L-Ser(α-D-Man(Ac)4)-OH 95%

Supplier: AFG Bioscience

Synonyms: N-α-(9-Fluorenylmethyloxycarbonyl)-O-β-(2,3,4,6-tetra-O-acetyl-α-D-mannopyranosyl)-L-serine

C51014-100MG C51014-250MG C51014-25MG
76619-304EA 778.81 CAD
76619-304 76619-306 76619-302
Fmoc-L-Ser(α-D-Man(Ac)4)-OH 95%
Fmoc-L-Ser(α-D-Man(Ac)4)-OH
Formula: C₃₂H₃₅NO₁₄
Storage Temperature: Freezer
MDL Number: MFCD05663656
CAS Number: 118358-80-8

Order Now

Specification Test Results

Purity Limit 95%
Appearance White powder


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR