Fmoc-L-Ser(a-D-Glc(Ac)4)-OH ≥95%

Supplier: AFG Bioscience

Synonyms: 2,3,4,6-Tetra-O-acetyl-a-D-glucopyranosyl-Fmoc-L-serine

C51013-100MG C51013-25MG C51013-50MG
76619-300EA 1978.02 CAD
76619-300 76619-296 76619-298
Fmoc-L-Ser(a-D-Glc(Ac)4)-OH ≥95%
Fmoc-L-Ser(a-D-Glc(Ac)4)-OH
Formula: C₃₂H₃₅NO₁₄
MW: 657.62 g/mol
Storage Temperature: Freezer
CAS Number: 337903-65-8

Order Now

Specification Test Results

Purity Limit > 95%
Appearance White Powder


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR