Fmoc-L-Ser (β-D-GalNAc(Ac)3)-OH ≥95%

Supplier: AFG Bioscience

C51006-100MG C51006-250MG C51006-25MG
76619-274EA 778.81 CAD
76619-274 76619-276 76619-272
Fmoc-L-Ser (β-D-GalNAc(Ac)3)-OH ≥95%
Fmoc-L-Ser (β-D-GalNAc(Ac)3)-OH
Formula: C₃₂H₃₆N₂O₁₃
Storage Temperature: Freezer
CAS Number: 1676104-71-4

Order Now

Specification Test Results

Purity Limit > 95%
Appearance White Powder


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR