Fmoc-L-Asn((Ac)4-β-D-Glc)-OH ≥95%

Supplier: AFG Bioscience

Synonyms: N-α-(fluorenylmethoxycarbonyl)-N-γ-(2,3,4,6-tetra-O-acetyl-β-D-glucopyranosyl)-L-asparagine

C51002-100MG C51002-250MG C51002-500MG
76619-260EA 389.41 CAD
76619-260 76619-262 76619-264
Fmoc-L-Asn((Ac)4-β-D-Glc)-OH ≥95%
Fmoc-L-Asn((Ac)4-β-D-Glc)-OH
Formula: C₃₃H₃₆N₂O₁₄
Storage Temperature: Freezer
CAS Number: 154395-64-9

Order Now

Specification Test Results

Purity Limit > 95%
Appearance White Powder


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR