2-amino-2,6-dideoxy-L-talose HCl salt ≥95%

Supplier: AFG BIOSCIENCE LLC

Synonyms: 6-deoxy-L-talosamine HCl salt

C50743-10MG C50743-5MG
76619-252EA 2136.25 CAD
76619-252 76619-250
2-amino-2,6-dideoxy-L-talose HCl salt ≥95%
2-amino-2,6-dideoxy-L-talose HCl salt
Formula: C₆ClH₁₄NO₄
MW: 199.63 g/mol
Storage Temperature: Refrigerator
CAS Number: 5867-12-9

Order Now

Specification Test Results

Purity Limit > 95%
Appearance white powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR