Methyl 2,3,6-tri-O-benzyl-α-D-glucopyranoside ≥95%

Supplier: AFG Bioscience

C50727-1G C50727-2G C50727-5G
76619-222EA 623.06 CAD
76619-222 76619-224 76619-226
Methyl 2,3,6-tri-O-benzyl-α-D-glucopyranoside ≥95%
Methyl 2,3,6-tri-O-benzyl-α-D-glucopyranoside
Formula: C₂₈H₃₂O₆
Storage Temperature: Refrigerator
MDL Number: MFCD04973355
CAS Number: 19488-48-3

Order Now

Specification Test Results

Purity Limit > 95%
Appearance White Powder


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR