2-Naphthyl-β-D-galactopyranoside hydrate ≥95%

Supplier: AFG Bioscience

C50726-1G C50726-5G
76619-218EA 316.47 CAD
76619-218 76619-220
2-Naphthyl-β-D-galactopyranoside hydrate ≥95%
2-Naphthyl-β-D-galactopyranoside hydrate
Formula: C₁₆H₁₈O₆
Storage Temperature: Refrigerator
MDL Number: MFCD00149268
CAS Number: 312693-81-5

Order Now

Specification Test Results

Purity Limit > 95%
Appearance White Powder


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR