2,3,4-Tribenzoyl-L-rhamnopyranose ≥95%
Supplier: AFG BIOSCIENCE LLC
Synonyms:
6-Deoxymannopyranose 2,3,4-tribenzoate
C50723-1G
C50723-5G
76619-210EA
155.76
CAD
76619-210
76619-212
2,3,4-Tribenzoyl-L-rhamnopyranose ≥95%
2,3,4-Tribenzoyl-L-rhamnopyranose
Formula:
C₂₇H₂₄O₈ MW: 476.47 g/mol Storage Temperature: Freezer |
CAS Number:
7494-44-2 |
Specification Test Results
Purity Limit | > 95% |
Appearance | Powder or Crystals |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...