4-Nitrophenyl hepta-O-acetyl-1-thio-β-lactoside 95%

Supplier: AFG BIOSCIENCE LLC

Synonyms: 4-Nitrophenyl 2,3,6-tri-O-acetyl-4-O-(2,3,4,6-tetra-O-acetyl-β-D-galactopyranosyl)-1-thio-β-D-glucopyranoside

C50718-100MG C50718-250MG
76619-202EA 870.33 CAD
76619-202 76619-204
4-Nitrophenyl hepta-O-acetyl-1-thio-β-lactoside 95%
4-Nitrophenyl hepta-O-acetyl-1-thio-β-lactoside
Formula: C₃₂H₃₉NO₁₉S
MW: 773.71 g/mol
Storage Temperature: Freezer
MDL Number: MFCD05664736
CAS Number: 78006-49-2

Order Now

Specification Test Results

Purity Limit 95%
Appearance White solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR