Methyl 2,3,4-tri-O-acetyl-6-O-trityl-α-D-glucopyranoside 95%

Supplier: AFG Bioscience

C50716-1G C50716-5G
76619-192EA 233.65 CAD
76619-192 76619-194
Methyl 2,3,4-tri-O-acetyl-6-O-trityl-α-D-glucopyranoside 95%
Methyl 2,3,4-tri-O-acetyl-6-O-trityl-α-D-glucopyranoside
Formula: C₃₂H₃₄O₉
Storage Temperature: Freezer
MDL Number: MFCD09038452
CAS Number: 18031-49-7

Order Now

Specification Test Results

Purity Limit 95%
Appearance White solid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR