Methyl 2,3,4-tri-O-benzyl-6-O-trityl-α-D-glucopyranoside 95%

Supplier: AFG Bioscience

C50715-1G C50715-5G
76619-188EA 233.65 CAD
76619-188 76619-190
Methyl 2,3,4-tri-O-benzyl-6-O-trityl-α-D-glucopyranoside 95%
Methyl 2,3,4-tri-O-benzyl-6-O-trityl-α-D-glucopyranoside
Formula: C₄₇H₄₆O₆
Storage Temperature: Freezer
MDL Number: MFCD09952842
CAS Number: 18685-19-3

Order Now

Specification Test Results

Purity Limit 95%
Appearance White solid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR