6-O-Trityl-N-acetyl-D-glucosamine 97%
Supplier: AFG Bioscience
C50712-1G
C50712-5G
76619-174EA
316.47
CAD
76619-174
76619-176
6-O-Trityl-N-acetyl-D-glucosamine 97%
6-O-Trityl-N-acetyl-D-glucosamine
Formula:
C₂₇H₂₉NO₆ MW: 463.52 g/mol Storage Temperature: Refrigerator |
CAS Number:
6959-53-1 |
Specification Test Results
Purity Limit | 97% |
Appearance | White Solid |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...