L-xylulose 95%

Supplier: AFG BIOSCIENCE LLC

Synonyms: L-threo-Pentulose

C50711-100MG C50711-250MG C50711-50MG
76619-170EA 632.96 CAD
76619-170 76619-172 76619-168
L-xylulose 95%
L-xylulose
Formula: C₅H₁₀O₅
MW: 150.13 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00064286
CAS Number: 527-50-4

Order Now

Specification Test Results

Purity Limit 95%
Appearance Syrup

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR