L-Ribulose 95%

Supplier: AFG Bioscience

C50709-250MG C50709-500MG
76619-160EA 395.6 CAD
76619-160 76619-162
L-Ribulose 95%
L-Ribulose
Formula: C₅H₁₀O₅
Storage Temperature: Refrigerator
MDL Number: MFCD00075347
CAS Number: 2042-27-5

Order Now

Specification Test Results

Purity Limit 95%
Appearance Syrup


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR