α-​Neuraminic acid, N-​acetyl-​2-​O-​(phenylmethyl)​-​, methyl ester, 4,​7,​8,​9-​tetraacetate ≥95%

Supplier: AFG BIOSCIENCE LLC

C50708-1G C50708-2G C50708-5G
76619-154EA 632.96 CAD
76619-154 76619-156 76619-158
α-​Neuraminic acid, N-​acetyl-​2-​O-​(phenylmethyl)​-​, methyl ester, 4,​7,​8,​9-​tetraacetate ≥95%
α-​Neuraminic acid, N-​acetyl-​2-​O-​(phenylmethyl)​-​, methyl ester, 4,​7,​8,​9-​tetraacetate
Formula: C₂₇H₃₅NO₁₃
MW: 581.57 g/mol
Storage Temperature: Refrigerator
CAS Number: 19342-75-7

Order Now

Specification Test Results

Purity Limit > 95%
Appearance White Powder


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR