2-Acetamidoglucal 95%
Supplier: AFG Bioscience
C50693-100MG
C50693-250MG
C50693-500MG
76619-128EA
389.41
CAD
76619-128
76619-130
76619-132
2-Acetamidoglucal 95%
2-Acetamidoglucal
Formula:
C₈H₁₃NO₅ Storage Temperature: Refrigerator |
CAS Number:
10293-59-1 |
Specification Test Results
Purity Limit | 95% |
Appearance | light yellow powder |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...