N-(2-Azidoacetyl)-neuraminic acid ≥95%

Supplier: AFG Bioscience

C51557-10MG C51557-2MG C51557-5MG
76619-586EA 2373.62 CAD
76619-586 76619-582 76619-584
N-(2-Azidoacetyl)-neuraminic acid ≥95%
N-(2-Azidoacetyl)-neuraminic acid
Formula: C₁₁H₁₈N₄O₉
Storage Temperature: Refrigerator
CAS Number: 775306-77-9

Order Now

Specification Test Results

Purity Limit > 95%
Appearance white powder


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR