Fmoc-L-Ser(β-D-Glc(Ac)4)-OH 95%

Supplier: AFG Bioscience

Synonyms: 2,3,4,6-Tetra-O-acetyl-β-D-glucopyranosyl-Fmoc-L-serine

C51012-100MG C51012-250MG C51012-25MG
76619-292EA 623.06 CAD
76619-292 76619-294 76619-290
Fmoc-L-Ser(β-D-Glc(Ac)4)-OH 95%
Fmoc-L-Ser(β-D-Glc(Ac)4)-OH
Formula: C₃₂H₃₅NO₁₄
MW: 657.62 g/mol
Storage Temperature: Freezer
MDL Number: MFCD15145006
CAS Number: 118358-38-6

Order Now

Specification Test Results

Purity Limit 95%
Appearance White powder


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR