n-Octyl-β-D-Glucopyranoside tetraacetate 98%

Supplier: AFG BIOSCIENCE LLC

Synonyms: β-​D-​Glucopyranoside, octyl, 2,​3,​4,​6-​tetraacetate, (2R,3R,4S,5R,6R)-2-(Acetoxymethyl)-6-(octyloxy)tetrahydro-2H-pyran-3,4,5-triyl triacetate

C50687-100G C50687-10G C50687-25G C50687-5G
76619-126EA 700.94 CAD
76619-126 76619-122 76619-124 76619-120
n-Octyl-β-D-Glucopyranoside tetraacetate 98%
n-Octyl-β-D-Glucopyranoside tetraacetate
Formula: C₂₂H₃₆O₁₀
MW: 460.515 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00192368
CAS Number: 38954-67-5

Order Now

Specification Test Results

Purity Limit 98%
Appearance White to off-white powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR