N-Acetyl-D-lactosamine ≥95%
Supplier: AFG Bioscience
Synonyms:
N-Acetyllactosamine, 2-(Acetylamino)-2-deoxy-4-O-β-D-galactopyranosyl-D-glucose, N-((2R,3R,4S,5R)-3,5,6-Trihydroxy-1-oxo-4-(((2S,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)hexan-2-yl)acetamide
C50359-100MG
C50359-250MG
C50359-50MG
76619-096EA
474.09
CAD
76619-096
76619-098
76619-094
N-Acetyl-D-lactosamine ≥95%
N-Acetyl-D-lactosamine
Formula:
C₁₄H₂₅NO₁₁ Storage Temperature: Freezer |
MDL Number:
MFCD00063785 CAS Number: 32181-59-2 |
Specification Test Results
Purity Limit | > 95% |
Appearance | White Powder |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...